trans-4-(Boc-amino)cyclohexanol ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl (trans-4-hydroxycyclohexyl)carbamate, trans-4-Aminocyclohexan-1-ol, N-BOC protected

073547-1G 073547-5G 073547-10G
102530-742EA 90.77 USD
102530-742 102530-744 102530-746
trans-4-(Boc-amino)cyclohexanol ≥95%
trans-4-(Boc-amino)cyclohexanol

Hazard: Irritant

Formula: C₁₁H₂₁NO₃
MDL Number: MFCD03844613
CAS Number: 111300-06-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR