Ethyl-5-chloro-4-methylisoxazole-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 5-chloro-4-methylisoxazole-3-carboxylate

073543-250MG 073543-1G
102530-724EA 826.15 USD
102530-724 102530-726
Ethyl-5-chloro-4-methylisoxazole-3-carboxylate ≥95%
Ethyl-5-chloro-4-methylisoxazole-3-carboxylate

Hazard: Irritant

Formula: C₇H₈ClNO₃
MW: 189.6 g/mol
MDL Number: MFCD11520431
CAS Number: 3356-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR