6-Bromo-5-ethoxypyridin-3-ylamine ≥95%

Supplier: Matrix Scientific

073542-250MG 073542-1G 073542-5G
102530-718EA 355.38 USD
102530-718 102530-720 102530-722
6-Bromo-5-ethoxypyridin-3-ylamine ≥95%
6-Bromo-5-ethoxypyridin-3-ylamine

Hazard: Irritant

Formula: C₇H₉BrN₂O
CAS Number: 1020253-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR