4-Nitro-o-phenylenediamine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Nitro-1,2-phenylenediamine, 4-Nitrobenzene-1,2-diamine

073540-100G 073540-500G
102530-710EA 60 USD
102530-710 102530-712
4-Nitro-o-phenylenediamine ≥95%
4-Nitro-o-phenylenediamine

Hazard: Irritant

Formula: C₆H₇N₃O₂
MW: 153.14 g/mol
Melting Pt: 199…203 °C
MDL Number: MFCD00007724
CAS Number: 99-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR