3-Amino-5-bromopyridin-2(1H)-one ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Amino-5-bromo-2-hydroxypyridine
073538-1G
073538-5G
102530-698EA
138.46
USD
102530-698
102530-700
3-Amino-5-bromopyridin-2(1H)-one ≥95%
3-Amino-5-bromopyridin-2(1H)-one
Hazard: Irritant
Formula:
C5H5BrN2O MW: 189.01 g/mol |
MDL Number:
MFCD03840438 CAS Number: 98786-86-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...