5-Nitropyridin-3-amine ≥95%

Supplier: Matrix Scientific

073534-250MG 073534-1G 073534-5G
102530-676EA 541.54 USD
102530-676 102530-678 102530-680
5-Nitropyridin-3-amine ≥95%
5-Nitropyridin-3-amine

Hazard: Irritant

Formula: C5H5N3O2
MDL Number: MFCD04114212
CAS Number: 934-59-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR