2-Fluoro-5-methylbenzaldehyde ≥95%

Supplier: Matrix Scientific

073533-1G 073533-5G 073533-10G
102530-670EA 38.46 USD
102530-670 102530-672 102530-674
2-Fluoro-5-methylbenzaldehyde ≥95%
2-Fluoro-5-methylbenzaldehyde

Hazard: Irritant/Air Sensitive

Formula: C₈H₇FO
Boiling Pt: 89°C/15mm
Density: 1.129
MDL Number: MFCD02093762
CAS Number: 93249-44-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR