4,7-Dimethoxy-1,10-phenanthroline ≥95%

Supplier: Matrix Scientific

073532-250MG 073532-1G 073532-5G
102530-664EA 129.23 USD
102530-664 102530-666 102530-668
4,7-Dimethoxy-1,10-phenanthroline ≥95%
4,7-Dimethoxy-1,10-phenanthroline

Hazard: Irritant

Formula: C₁₄H₁₂N₂O₂
MW: 240.2573 g/mol
MDL Number: MFCD00233883
CAS Number: 92149-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR