5-Bromo-3-cyanoindole ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-1H-indole-3-carbonitrile

073530-1G 073530-5G 073530-10G
102530-656EA 640 USD
102530-656 102530-658 102530-660
5-Bromo-3-cyanoindole ≥95%
5-Bromo-3-cyanoindole

Hazard: Irritant

Formula: C₉H₅BrN₂
MW: 221.0500 g/mol
MDL Number: MFCD04127133
CAS Number: 90271-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR