3-Acetyl-2-bromopyridine ≥95%

Supplier: Matrix Scientific

073528-250MG 073528-1G 073528-5G
102530-644EA 427.69 USD
102530-644 102530-646 102530-648
3-Acetyl-2-bromopyridine ≥95%
3-Acetyl-2-bromopyridine

Hazard: Irritant

Formula: C₇H₆BrNO
MW: 200.0300 g/mol
MDL Number: MFCD04114370
CAS Number: 84199-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR