3-Iodopyridine-2-carbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Iodopicolinonitrile, 2-Cyano-3-iodopyridine

073527-250MG 073527-1G 073527-5G
102530-638EA 498.46 USD
102530-638 102530-640 102530-642
3-Iodopyridine-2-carbonitrile ≥95%
3-Iodopyridine-2-carbonitrile

Hazard: Irritant

Formula: C₆H₃IN₂
MW: 230.01 g/mol
MDL Number: MFCD09037451
CAS Number: 827616-52-4
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR