5-Amino-2-chlorobenzothiazole ≥95%

Supplier: Matrix Scientific

073526-250MG 073526-1G 073526-5G
102530-632EA 427.69 USD
102530-632 102530-634 102530-636
5-Amino-2-chlorobenzothiazole ≥95%
5-Amino-2-chlorobenzothiazole

Hazard: Irritant

Formula: C₇H₅ClN₂S
MW: 184.6460 g/mol
MDL Number: MFCD11040282
CAS Number: 80945-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR