1-N-Boc-4-(4-bromophenyl)piperidine ≥95%

Supplier: Matrix Scientific

Synonyms: 1-N-Boc-4-(4-Bromophenyl)piperidine

073522-1G 073522-5G
102530-614EA 315.38 USD
102530-614 102530-616
1-N-Boc-4-(4-bromophenyl)piperidine ≥95%
1-N-Boc-4-(4-bromophenyl)piperidine

Hazard: Irritant

Formula: C₁₆H₂₂BrNO₂
MW: 340.2600 g/mol
MDL Number: MFCD09878774
CAS Number: 769944-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR