4-Bromo-2-nitrophenol ≥95%

Supplier: Matrix Scientific

073521-100G 073521-25G
102530-612EA 230.77 USD
102530-612 102833-634
4-Bromo-2-nitrophenol ≥95%
4-Bromo-2-nitrophenol

Hazard: Irritant

Formula: C₆H₄BrNO₃
MDL Number: MFCD00082540
CAS Number: 7693-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR