Ethyl-5-formyl-1H-pyrrole-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 5-formyl-1H-pyrrole-2-carboxylate

073517-250MG 073517-1G
102530-586EA 267.69 USD
102530-586 102530-588
Ethyl-5-formyl-1H-pyrrole-2-carboxylate ≥95%
Ethyl-5-formyl-1H-pyrrole-2-carboxylate

Hazard: Irritant

Formula: C₈H₉NO₃
MW: 167.1600 g/mol
MDL Number: MFCD00159462
CAS Number: 7126-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR