1-Benzyl-4-cyano-4-phenylpiperidine hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
1-Benzyl-4-phenylpiperidine-4-carbonitrile hydrochloride
073516-10G
073516-100G
102530-582EA
64.62
USD
102530-582
102530-584
1-Benzyl-4-cyano-4-phenylpiperidine hydrochloride ≥95%
1-Benzyl-4-cyano-4-phenylpiperidine hydrochloride
Hazard: Irritant
Formula:
C₁₉H₂₁ClN₂ MW: 312.85 g/mol |
MDL Number:
MFCD00035361 CAS Number: 71258-18-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...