Imidazo[1,5-a]pyridine-7-carboxylic acid ≥95%

Supplier: Matrix Scientific

073514-250MG 073514-1G 073514-5G
102530-570EA 350.81 USD
102530-570 102530-572 102530-574
Imidazo[1,5-a]pyridine-7-carboxylic acid ≥95%
Imidazo[1,5-a]pyridine-7-carboxylic acid

Hazard: Irritant

Formula: C₈H₆N₂O₂
MDL Number: MFCD11870763
CAS Number: 588720-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR