2-Amino-6-phenylpyrimidin-4(3H)-one ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Amino-6-phenyl-4(1H)-pyrimidinone, 6-Phenylisocytosine
073509-1G
073509-5G
073509-10G
102530-544EA
249.23
USD
102530-544
102530-546
102530-548
2-Amino-6-phenylpyrimidin-4(3H)-one ≥95%
2-Amino-6-phenylpyrimidin-4(3H)-one
Hazard: Irritant
Formula:
C₁₀H₉N₃O MW: 187.2000 g/mol |
MDL Number:
MFCD00466412 CAS Number: 56741-94-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...