5-Bromo-1-isoindolinone ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-2,3-dihydro-1H-isoindol-1-one, 5-Bromoisoindolin-1-one

073506-1G 073506-5G
102530-532EA 75.38 USD
102530-532 102530-534
5-Bromo-1-isoindolinone ≥95%
5-Bromo-1-isoindolinone

Hazard: Irritant/Light Sensitive

Formula: C₈H₆BrNO
MW: 212.04 g/mol
MDL Number: MFCD09701292
CAS Number: 552330-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR