1-Methylpiperidin-3-one hydrochloride ≥95%

Supplier: Matrix Scientific

073504-1G 073504-5G
102530-522EA 289.23 USD
102530-522 102530-524
1-Methylpiperidin-3-one hydrochloride ≥95%
1-Methylpiperidin-3-one hydrochloride

Hazard: Irritant

Formula: C₆H₁₂ClNO
MDL Number: MFCD18432588
CAS Number: 41511-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR