2-Amino-4-methoxybenzothiazole ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Methoxybenzothiazol-2-ylamine

073503-1G 073503-5G 073503-10G
102530-514EA 143.08 USD
102530-514 102530-516 102530-518
2-Amino-4-methoxybenzothiazole ≥95%
2-Amino-4-methoxybenzothiazole

Hazard: Irritant

Formula: C₈H₈N₂OS
MW: 180.23 g/mol
MDL Number: MFCD00005792
CAS Number: 5464-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR