Methyl-7-bromoheptanoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 7-bromoheptanoate

073499-1G 073499-5G 073499-10G
102530-492EA 29.23 USD
102530-492 102530-494 102530-496
Methyl-7-bromoheptanoate ≥95%
Methyl-7-bromoheptanoate

Hazard: Irritant

Formula: C₈H₁₅BrO₂
Density: 1.257
MDL Number: MFCD02258672
CAS Number: 54049-24-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR