2-Chlorothiophene-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

073497-1G 073497-5G
102530-482EA 444.62 USD
102530-482 102530-484
2-Chlorothiophene-3-carboxylic acid ≥95%
2-Chlorothiophene-3-carboxylic acid

Hazard: Irritant

Formula: C₅H₃ClO₂S
MDL Number: MFCD00098423
CAS Number: 53935-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR