1-Aminocyclopentanecarboxylic acid ≥95%

Supplier: Matrix Scientific

073495-5G 073495-100G
102530-468EA 53.85 USD
102530-468 102530-472
1-Aminocyclopentanecarboxylic acid ≥95%
1-Aminocyclopentanecarboxylic acid

Hazard: Irritant

Formula: C₆H₁₁NO₂
MDL Number: MFCD00001381
CAS Number: 52-52-8
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR