5-Amino-2,4-dichloropyrimidine ≥95%

Supplier: Matrix Scientific

Synonyms: 2,4-Dichloropyrimidin-5-amine

073493-5G 073493-25G
102530-458EA 67.69 USD
102530-458 102833-632
5-Amino-2,4-dichloropyrimidine ≥95%
5-Amino-2,4-dichloropyrimidine

Hazard: Irritant

Formula: C₄H₃Cl₂N₃
MW: 164.00 g/mol
MDL Number: MFCD05662684
CAS Number: 5177-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR