(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one ≥95%

Supplier: Matrix Scientific

Synonyms: 2-azabicyclo[2.2.1]hept-5-en-3-one

073490-100G 073490-25G
102530-446EA 192.31 USD
102530-446 102551-846
(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one ≥95%
(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one

Hazard: Irritant

Formula: C₆H₇NO
MDL Number: MFCD00213364
CAS Number: 49805-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR