1-Isoquinolinol ≥95%

Supplier: Matrix Scientific

073488-1G 073488-5G 073488-10G
102530-434EA 49.23 USD
102530-434 102530-436 102530-438
1-Isoquinolinol ≥95%
1-Isoquinolinol

Hazard: Irritant

Formula: C₉H₇NO
MW: 145.16 g/mol
MDL Number: MFCD00006899
CAS Number: 491-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR