(3R,4R)-1-Benzyl-N-4-dimethylpiperidin-3-amine ≥95%

Supplier: Matrix Scientific

Synonyms: (3R,4R)-1-Benzyl-N,4-dimethylpiperidin-3-amine

073486-250MG 073486-1G
102530-422EA 380 USD
102530-422 102530-424
(3R,4R)-1-Benzyl-N-4-dimethylpiperidin-3-amine ≥95%
(3R,4R)-1-Benzyl-N-4-dimethylpiperidin-3-amine

Hazard: Irritant

Formula: C₁₄H₂₂N₂
MW: 218.3379 g/mol
MDL Number: MFCD09475548
CAS Number: 477600-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR