(S)-3-Hydroxypiperidine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-3-Piperidinol hydrochloride

073484-5G 073484-25G
102530-414EA 107.69 USD
102530-414 102833-630
(S)-3-Hydroxypiperidine hydrochloride ≥95%
(S)-3-Hydroxypiperidine hydrochloride

Hazard: Irritant

Formula: C₅H₁₂ClNO
MW: 137.6100 g/mol
MDL Number: MFCD03426348
CAS Number: 475058-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR