(5-Bromo-3-pyridinyl)methanol ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Bromo-5-(hydroxymethyl)pyridine, 5-Bromo-3-pyridinemethanol
073479-1G
073479-5G
102530-384EA
46.15
USD
102530-384
102530-386
(5-Bromo-3-pyridinyl)methanol ≥95%
(5-Bromo-3-pyridinyl)methanol
Hazard: Irritant
Formula:
C₆H₆BrNO MW: 188.0200 g/mol |
MDL Number:
MFCD03265757 CAS Number: 37669-64-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...