2-(1-Aminocyclohexyl)acetic acid hydrochloride ≥95%

Supplier: Matrix Scientific

073478-250MG 073478-1G 073478-5G
102530-378EA 355.38 USD
102530-378 102530-380 102530-382
2-(1-Aminocyclohexyl)acetic acid hydrochloride ≥95%
2-(1-Aminocyclohexyl)acetic acid hydrochloride

Hazard: Irritant

Formula: C₈H₁₆ClNO₂
MW: 193.6700 g/mol
MDL Number: MFCD06411715
CAS Number: 37631-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR