6-Methylpyridine-2,4-diol ≥95%

Supplier: Matrix Scientific

073476-1G 073476-5G 073476-10G
102530-368EA 15.38 USD
102530-368 102530-370 102530-372
6-Methylpyridine-2,4-diol ≥95%
6-Methylpyridine-2,4-diol

Hazard: Irritant

Formula: C₆H₇NO₂
MW: 125.13 g/mol
MDL Number: MFCD01860849
CAS Number: 158152-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR