2-(Chloromethyl)pyrimidine hydrochloride ≥95%

Supplier: Matrix Scientific

073474-1G 073474-5G
102530-356EA 190.77 USD
102530-356 102530-358
2-(Chloromethyl)pyrimidine hydrochloride ≥95%
2-(Chloromethyl)pyrimidine hydrochloride

Hazard: Irritant/Refrigerate

Formula: C₅H₆Cl₂N₂
MW: 165.02 g/mol
Melting Pt: 138…140 °C
MDL Number: MFCD09834372
CAS Number: 936643-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR