2-Chloro-3-fluoro-6-methylpyridine ≥95%

Supplier: Matrix Scientific

073472-1G 073472-5G
102530-346EA 533.85 USD
102530-346 102530-348
2-Chloro-3-fluoro-6-methylpyridine ≥95%
2-Chloro-3-fluoro-6-methylpyridine

Hazard: Irritant

Formula: C₆H₅ClFN
MW: 145.56 g/mol
MDL Number: MFCD03095216
CAS Number: 374633-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR