5-Chlorothiophene-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

073470-1G 073470-5G
102530-338EA 136.92 USD
102530-338 102530-340
5-Chlorothiophene-3-carboxylic acid ≥95%
5-Chlorothiophene-3-carboxylic acid

Hazard: Irritant

Formula: C5H3ClO2S
MDL Number: MFCD00082877
CAS Number: 36157-42-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR