2-Bromo-3,5-dichloroisonicotinic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-3,5-dichloropyridine-4-carboxylic acid

073465-1G 073465-5G
102530-312EA 115.38 USD
102530-312 102530-314
2-Bromo-3,5-dichloroisonicotinic acid ≥95%
2-Bromo-3,5-dichloroisonicotinic acid

Hazard: Irritant

Formula: C₆H₂BrCl₂NO₂
MW: 270.90 g/mol
MDL Number: MFCD03411637
CAS Number: 343781-56-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR