Ethyl-5-chloro-3-methylisoxazole-4-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 5-chloro-3-methylisoxazole-4-carboxylate
073464-250MG
073464-1G
073464-5G
102530-306EA
640
USD
102530-306
102530-308
102530-310
Ethyl-5-chloro-3-methylisoxazole-4-carboxylate ≥95%
Ethyl-5-chloro-3-methylisoxazole-4-carboxylate
Hazard: Irritant
Formula:
C₇H₈ClNO₃ MW: 189.6000 g/mol |
MDL Number:
MFCD11557159 CAS Number: 3356-94-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...