Isopropyl-6-chloropyridazine-3-carboxylate ≥95%

Supplier: Matrix Scientific

073460-250MG 073460-1G 073460-5G
102530-282EA 684.62 USD
102530-282 102530-284 102530-286
Isopropyl-6-chloropyridazine-3-carboxylate ≥95%
Isopropyl-6-chloropyridazine-3-carboxylate

Hazard: Irritant

Formula: C₈H₉ClN₂O₂
CAS Number: 321946-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR