3-Methoxyanthranilic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-m-anisic acid, 3-Methoxyanthranilic acid, o-Anisidine-6-carboxylic acid, 2-Amino-3-methoxybenzoic acid

073459-1G 073459-5G
102530-278EA 64.62 USD
102530-278 102530-280
3-Methoxyanthranilic acid ≥95%
3-Methoxyanthranilic acid

Hazard: Irritant

Formula: C₈H₉NO₃
MW: 167.16 g/mol
MDL Number: MFCD00075178
CAS Number: 3177-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR