(S)-tert-Butylmethyl(piperidin-3-yl)carbamate ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-tert-Butyl methyl(piperidin-3-yl)carbamate

073457-250MG 073457-1G 073457-5G
102530-266EA 136.92 USD
102530-266 102530-268 102530-270
(S)-tert-Butylmethyl(piperidin-3-yl)carbamate ≥95%
(S)-tert-Butylmethyl(piperidin-3-yl)carbamate

Hazard: Irritant

Formula: C₁₁H₂₂N₂O₂
MDL Number: MFCD09878597
CAS Number: 309962-63-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR