1-Ethynyl-3-nitrobenzene ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Nitrophenylacetylene, 1-Ethynyl-3-nitrobenzene, (3-Nitrophenyl)ethyne, (3-Nitrophenyl)acetylene

073455-1G 073455-5G
102530-256EA 166.15 USD
102530-256 102530-258
1-Ethynyl-3-nitrobenzene ≥95%
1-Ethynyl-3-nitrobenzene

Hazard: Irritant

Formula: C₈H₅NO₂
MW: 147.13
MDL Number: MFCD02181150
CAS Number: 3034-94-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR