tert-Butyl-3-(hydroxymethyl)piperazine-1-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
1-(tert-Butoxycarbonyl)-3-(hydroxymethyl)piperazine, 1-Boc-3-(hydroxymethyl)piperazine, 3-(Hydroxymethyl)piperazine-1-carboxylic acid tert-Butyl ester, tert-Butyl 3-(Hydroxymethyl)piperazine-1-carboxylate, tert-Butyl 3-(hydroxymethyl)piperazine-1-carboxylate
073454-1G
073454-5G
102530-252EA
75.38
USD
102530-252
102530-254
tert-Butyl-3-(hydroxymethyl)piperazine-1-carboxylate ≥95%
tert-Butyl-3-(hydroxymethyl)piperazine-1-carboxylate
Hazard: Irritant
Formula:
C₁₀H₂₀N₂O₃ MW: 216.2774 g/mol |
MDL Number:
MFCD04115304 CAS Number: 301673-16-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...