5-Methoxy-2-pyridinecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Methoxypicolinic acid

073452-1G 073452-5G
102530-244EA 98.46 USD
102530-244 102530-246
5-Methoxy-2-pyridinecarboxylic acid ≥95%
5-Methoxy-2-pyridinecarboxylic acid

Hazard: Irritant

Formula: C7H7NO3
MW: 153.1400 g/mol
MDL Number: MFCD06800961
CAS Number: 29082-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR