2,6-Dichloro-3-nitropyridin-4-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Amino-2,6-dichloro-3-nitropyridine

073451-1G 073451-5G
102530-238EA 123.08 USD
102530-238 102530-240
2,6-Dichloro-3-nitropyridin-4-amine ≥95%
2,6-Dichloro-3-nitropyridin-4-amine

Hazard: Irritant

Formula: C₅H₃Cl₂N₃O₂
MW: 208.0022 g/mol
MDL Number: MFCD03265754
CAS Number: 2897-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR