4,5-Dichloro-2-fluorobenzoic acid ≥95%

Supplier: Matrix Scientific

073450-1G 073450-5G 073450-25G
102530-232EA 92.31 USD
102530-232 102530-234 102551-842
4,5-Dichloro-2-fluorobenzoic acid ≥95%
4,5-Dichloro-2-fluorobenzoic acid

Hazard: Irritant

Formula: C₇H₃Cl₂FO₂
MW: 209.0019 g/mol
MDL Number: MFCD00672988
CAS Number: 289039-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR