1-Methyl-1H-benzimidazole-5-carboxaldehyde ≥95%

Supplier: Matrix Scientific

073448-250MG 073448-1G
102530-222EA 967.69 USD
102530-222 102530-224
1-Methyl-1H-benzimidazole-5-carboxaldehyde ≥95%
1-Methyl-1H-benzimidazole-5-carboxaldehyde

Hazard: Irritant

Formula: C₉H₈N₂O
MDL Number: MFCD09261052
CAS Number: 279226-70-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR