(1-Pyridin-4-yl)piperidine ≥95%

Supplier: Matrix Scientific

073446-1G 073446-5G
102530-212EA 640 USD
102530-212 102530-214
(1-Pyridin-4-yl)piperidine ≥95%
(1-Pyridin-4-yl)piperidine

Hazard: Irritant

Formula: C₁₀H₁₄N₂
MW: 162.2300 g/mol
MDL Number: MFCD00006476
CAS Number: 2767-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR