2,3-Diamino-5-methylpyridine ≥95%

Supplier: Matrix Scientific

073444-1G 073444-5G
102530-202EA 449.23 USD
102530-202 102530-204
2,3-Diamino-5-methylpyridine ≥95%
2,3-Diamino-5-methylpyridine

Hazard: Irritant

Formula: C₆H₉N₃
CAS Number: 24638-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR