Methyl-3-methoxypicolinate ≥95%

Supplier: Matrix Scientific

Synonyms: methyl 3-methoxypyridine-2-carboxylate

073443-250MG 073443-1G
102530-196EA 684.62 USD
102530-196 102530-198
Methyl-3-methoxypicolinate ≥95%
Methyl 3-methoxypicolinate

Hazard: Irritant

Formula: C₈H₉NO₃
MW: 167.1600 g/mol
MDL Number: MFCD06412569
CAS Number: 24059-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR