4-Oxazolecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: Oxazole-4-carboxylic acid

073439-1G 073439-5G 073439-10G
102530-172EA 63.08 USD
102530-172 102530-174 102530-176
4-Oxazolecarboxylic acid ≥95%
4-Oxazolecarboxylic acid

Hazard: Irritant

Formula: C₄H₃NO₃
MW: 113.0715 g/mol
MDL Number: MFCD06797151
CAS Number: 23012-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR